(3E)-2-(4-Chlorophenyl)-3-(methoxyimino)propanenitrile, tech

Lieferant: Apollo Scientific

OR32860-1G
APOSOR32860-1GEA 1570 EUR
APOSOR32860-1G
(3E)-2-(4-Chlorophenyl)-3-(methoxyimino)propanenitrile, tech
(3E)-2-(4-Chlorophenyl)-3-(methoxyimino)propanenitrile
Formel: C₁₀H₉ClN₂O
Molecular Weight: 208,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975574
CAS-Nummer: 1013061-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR