Methyl (2E)-3-(1-benzofuran-2-yl)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32861-1G
APOSOR32861-1GEA 1570 EUR
APOSOR32861-1G
Methyl (2E)-3-(1-benzofuran-2-yl)prop-2-enoate, tech
Methyl (2E)-3-(1-benzofuran-2-yl)prop-2-enoate
Formel: C₁₂H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00231969
CAS-Nummer: 256439-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR