Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32862-1G
APOSOR32862-1GEA 1570 EUR
APOSOR32862-1G
Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate, tech
Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496183
CAS-Nummer: 956764-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR