(2E)-3-[(3,5-Dichlorophenyl)amino]prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR32863-1G
APOSOR32863-1GEA 1570 EUR
APOSOR32863-1G
(2E)-3-[(3,5-Dichlorophenyl)amino]prop-2-enenitrile, tech
(2E)-3-[(3,5-Dichlorophenyl)amino]prop-2-enenitrile
Formel: C₉H₆Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00663724
CAS-Nummer: 338959-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR