(3E)-3-{[(2,4-Dichlorophenyl)methoxy]imino}propanenitrile, tech

Lieferant: Apollo Scientific

OR32865-1G
APOSOR32865-1GEA 1570 EUR
APOSOR32865-1G
(3E)-3-{[(2,4-Dichlorophenyl)methoxy]imino}propanenitrile, tech
(3E)-3-{[(2,4-Dichlorophenyl)methoxy]imino}propanenitrile
Formel: C₁₀H₈Cl₂N₂O
Molecular Weight: 243,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663727
CAS-Nummer: 338959-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR