4-Chloro-5-(methylamino)-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, tech
Lieferant: Apollo Scientific
OR32871-1G
APOSOR32871-1GEA
1570
EUR
APOSOR32871-1G
4-Chloro-5-(methylamino)-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, tech
4-Chloro-5-(methylamino)-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one
Formel:
C₁₂H₁₂ClN₃O Molecular Weight: 249,696 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02083234 CAS-Nummer: 37627-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...