9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one, tech

Lieferant: Apollo Scientific

OR32875-1G
APOSOR32875-1GEA 1570 EUR
APOSOR32875-1G
9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one, tech
9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one
Formel: C₁₅H₁₆N₂O
Molecular Weight: 240,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00445032
CAS-Nummer: 130186-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR