3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione, tech

Lieferant: Apollo Scientific

OR32877-1G
APOSOR32877-1GEA 1570 EUR
APOSOR32877-1G
3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione, tech
3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00101089
CAS-Nummer: 24756-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR