(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32878-1G
APOSOR32878-1GEA 1570 EUR
APOSOR32878-1G
(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
(2E)-3-(Furan-2-yl)-1-(pyridin-2-yl)prop-2-en-1-one
Formel: C₁₂H₉NO₂
Molecular Weight: 199,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315211
CAS-Nummer: 18551-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR