(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32879-1G
APOSOR32879-1GEA 1570 EUR
APOSOR32879-1G
(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one, tech
(2E)-1,3-Bis(furan-2-yl)prop-2-en-1-one
Formel: C₁₁H₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00020927
CAS-Nummer: 42811-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR