4-Aminopyrimidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyrimidin-4-amine hydrochloride, 4-Amino-1,3-diazine hydrochloride

OR3288-100MG
APOSOR3288-100MGEA 21.9 EUR
APOSOR3288-100MG
4-Aminopyrimidine hydrochloride
4-Aminopyrimidine hydrochloride
Formel: C₄H₅N₃·ClH
Molecular Weight: 131,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702435
CAS-Nummer: 174500-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR