(2E)-3-(Furan-2-yl)-1-(pyridin-3-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32880-1G
APOSOR32880-1GEA 1570 EUR
APOSOR32880-1G
(2E)-3-(Furan-2-yl)-1-(pyridin-3-yl)prop-2-en-1-one, tech
(2E)-3-(Furan-2-yl)-1-(pyridin-3-yl)prop-2-en-1-one
Formel: C₁₂H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00224733
CAS-Nummer: 18461-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR