(2E)-3-(Furan-3-yl)-1-phenylprop-2-en-1-one, tech
Lieferant: Apollo Scientific
OR32881-1G
APOSOR32881-1GEA
1570
EUR
APOSOR32881-1G
(2E)-3-(Furan-3-yl)-1-phenylprop-2-en-1-one, tech
(2E)-3-(Furan-3-yl)-1-phenylprop-2-en-1-one
Formel:
C₁₃H₁₀O₂ Molecular Weight: 198,217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00449604 CAS-Nummer: 343571-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...