Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate 95%

Lieferant: Apollo Scientific

OR32882-1G
APOSOR32882-1GEA 1070 EUR
APOSOR32882-1G
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate 95%
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316055
CAS-Nummer: 476335-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR