4-Chloro-N-(2-hydroxy-1-phenylethyl)benzamide, tech

Lieferant: Apollo Scientific

OR32884-1G
APOSOR32884-1GEA 1570 EUR
APOSOR32884-1G
4-Chloro-N-(2-hydroxy-1-phenylethyl)benzamide, tech
4-Chloro-N-(2-hydroxy-1-phenylethyl)benzamide
Formel: C₁₅H₁₄ClNO₂
Molecular Weight: 275,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316059
CAS-Nummer: 338963-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR