Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate, tech

Lieferant: Apollo Scientific

OR32885-1G
APOSOR32885-1GEA 0 EUR
APOSOR32885-1G
Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate, tech
Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate
Formel: C₁₃H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00973916
CAS-Nummer: 86364-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR