[5-(4-Chlorophenyl)-1,2-oxazol-3-yl]methanol 95%

Lieferant: Apollo Scientific

OR32886-1G
APOSOR32886-1GEA 671 EUR
APOSOR32886-1G
[5-(4-Chlorophenyl)-1,2-oxazol-3-yl]methanol 95%
[5-(4-Chlorophenyl)-1,2-oxazol-3-yl]methanol
Formel: C₁₀H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01444076
CAS-Nummer: 81282-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR