[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR3289-250MG
APOSOR3289-250MGEA
37.7
EUR
APOSOR3289-250MG
[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
Formel:
C₁₂H₁₂N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665860 CAS-Nummer: 959584-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...