Nicotinaldehydoxim 95%

Lieferant: Apollo Scientific

Synonyme: 3-Pyridinealdoxime, Pyridine-3-carboxaldoxime, Pyridine-3-aldoxime

OR32892-25G
APOSOR32892-25GEA 126 EUR
APOSOR32892-25G
Nicotinaldehydoxim 95%
Nicotinaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006409
CAS-Nummer: 1193-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR