1-[(4-Methylphenyl)methoxy]-1H-imidazole-2-carbaldehyde, tech
Lieferant: Apollo Scientific
OR32893-1G
APOSOR32893-1GEA
1570
EUR
APOSOR32893-1G
1-[(4-Methylphenyl)methoxy]-1H-imidazole-2-carbaldehyde, tech
1-[(4-Methylphenyl)methoxy]-1H-imidazole-2-carbaldehyde
Formel:
C₁₂H₁₂N₂O₂ Molecular Weight: 216,236 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871523 CAS-Nummer: 478050-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...