1-(2-Phenylethoxy)-1H-imidazole-2-carbaldehyde, tech

Lieferant: Apollo Scientific

OR32894-1G
APOSOR32894-1GEA 1570 EUR
APOSOR32894-1G
1-(2-Phenylethoxy)-1H-imidazole-2-carbaldehyde, tech
1-(2-Phenylethoxy)-1H-imidazole-2-carbaldehyde
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871524
CAS-Nummer: 478050-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR