6,8-Dimethyl-[1,2,4]triazolo[4,3-b]pyridazine 95%
Lieferant: Apollo Scientific
OR32898-1G
APOSOR32898-1GEA
671
EUR
APOSOR32898-1G
6,8-Dimethyl-[1,2,4]triazolo[4,3-b]pyridazine 95%
6,8-Dimethyl-[1,2,4]triazolo[4,3-b]pyridazine
Formel:
C₇H₈N₄ Molecular Weight: 148,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00457973 CAS-Nummer: 18591-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...