4-(2,5-Dimethyl-1H-pyrrol-1-yl)-4H-1,2,4-triazole, tech

Lieferant: Apollo Scientific

OR32899-1G
APOSOR32899-1GEA 1570 EUR
APOSOR32899-1G
4-(2,5-Dimethyl-1H-pyrrol-1-yl)-4H-1,2,4-triazole, tech
4-(2,5-Dimethyl-1H-pyrrol-1-yl)-4H-1,2,4-triazole
Formel: C₈H₁₀N₄
Molecular Weight: 162.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187165
CAS-Nummer: 54931-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR