N-(2-Chloroethyl)-2-phenylacetamide, tech

Lieferant: Apollo Scientific

OR32901-1G
APOSOR32901-1GEA 1570 EUR
APOSOR32901-1G
N-(2-Chloroethyl)-2-phenylacetamide, tech
N-(2-Chloroethyl)-2-phenylacetamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186882
CAS-Nummer: 23547-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR