Methyl (2E)-3-[(E)-4-methylbenzoyloxy]prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32902-1G
APOSOR32902-1GEA 1570 EUR
APOSOR32902-1G
Methyl (2E)-3-[(E)-4-methylbenzoyloxy]prop-2-enoate, tech
Methyl (2E)-3-[(E)-4-methylbenzoyloxy]prop-2-enoate
Formel: C₁₂H₁₂O₄
Molecular Weight: 220,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187308
CAS-Nummer: 551931-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR