1-Benzyl-N'-[(1E)-(dimethylamino)methylidene]piperidine-4-carbohydrazide, tech

Lieferant: Apollo Scientific

OR32903-1G
APOSOR32903-1GEA 1570 EUR
APOSOR32903-1G
1-Benzyl-N'-[(1E)-(dimethylamino)methylidene]piperidine-4-carbohydrazide, tech
1-Benzyl-N'-[(1E)-(dimethylamino)methylidene]piperidine-4-carbohydrazide
Formel: C₁₆H₂₄N₄O
Molecular Weight: 288,388 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187020
CAS-Nummer: 672949-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR