1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR32906-1G
APOSOR32906-1GEA 1570 EUR
APOSOR32906-1G
1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one
Formel: C₁₁H₈ClNOS
Molecular Weight: 237,705 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187036
CAS-Nummer: 672950-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR