1-Acetyl-2,2,6,6-tetramethylpiperidin-4-one, tech

Lieferant: Apollo Scientific

OR32908-1G
APOSOR32908-1GEA 1570 EUR
APOSOR32908-1G
1-Acetyl-2,2,6,6-tetramethylpiperidin-4-one, tech
1-Acetyl-2,2,6,6-tetramethylpiperidin-4-one
Formel: C₁₁H₁₉NO₂
Molecular Weight: 197,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571209
CAS-Nummer: 52326-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR