Ethyl 4-chloro-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%

Lieferant: Apollo Scientific

OR32911-1G
APOSOR32911-1GEA 874 EUR
APOSOR32911-1G
Ethyl 4-chloro-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%
Ethyl 4-chloro-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Formel: C₁₀H₁₀ClN₃O₂
Molecular Weight: 239,658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012939
CAS-Nummer: 37801-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR