3-{6-Chloroimidazo[1,2-a]pyridin-2-yl}aniline, tech

Lieferant: Apollo Scientific

OR32917-1G
APOSOR32917-1GEA 1570 EUR
APOSOR32917-1G
3-{6-Chloroimidazo[1,2-a]pyridin-2-yl}aniline, tech
3-{6-Chloroimidazo[1,2-a]pyridin-2-yl}aniline
Formel: C₁₃H₁₀ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03012831
CAS-Nummer: 439110-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR