N-tert-Butyl-4-methylpiperazine-1-carbothioamide, tech
Lieferant: Apollo Scientific
OR32918-1G
APOSOR32918-1GEA
1570
EUR
APOSOR32918-1G
N-tert-Butyl-4-methylpiperazine-1-carbothioamide, tech
N-tert-Butyl-4-methylpiperazine-1-carbothioamide
Formel:
C₁₀H₂₁N₃S Molecular Weight: 215,359 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03102524 CAS-Nummer: 692287-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...