1-[5-(4-Methylpiperazin-1-yl)thiophen-2-yl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR32919-1G
APOSOR32919-1GEA 1570 EUR
APOSOR32919-1G
1-[5-(4-Methylpiperazin-1-yl)thiophen-2-yl]ethan-1-one, tech
1-[5-(4-Methylpiperazin-1-yl)thiophen-2-yl]ethan-1-one
Formel: C₁₁H₁₆N₂OS
Molecular Weight: 224,323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617419
CAS-Nummer: 685107-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR