6-Chlorimidazo[1,2-b]pyridazin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3292-25G
OR3292-5G
OR3292-100G
APOSOR3292-25GEA
78.3
EUR
APOSOR3292-25G
APOSOR3292-5G
APOSOR3292-100G
6-Chlorimidazo[1,2-b]pyridazin 98%
6-Chlorimidazo[1,2-b]pyridazin
Formel:
C₆H₄ClN₃ Schmelzpunkt: 115,5…117.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07778345 CAS-Nummer: 6775-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...