(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate, tech

Lieferant: Apollo Scientific

OR32920-1G
APOSOR32920-1GEA 1570 EUR
APOSOR32920-1G
(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate, tech
(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate
Formel: C₇H₆Cl₂N₂O₃
Molecular Weight: 237,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305724
CAS-Nummer: 97137-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR