Methyl 3-[(E)-[(dimethylamino)methylidene]amino]-1-benzofuran-2-carboxylate, tech

Lieferant: Apollo Scientific

OR32922-1G
APOSOR32922-1GEA 1570 EUR
APOSOR32922-1G
Methyl 3-[(E)-[(dimethylamino)methylidene]amino]-1-benzofuran-2-carboxylate, tech
Methyl 3-[(E)-[(dimethylamino)methylidene]amino]-1-benzofuran-2-carboxylate
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246.262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787753
CAS-Nummer: 866039-69-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR