4-Chloro-3-methanehydrazonoylquinoline, tech

Lieferant: Apollo Scientific

OR32924-1G
APOSOR32924-1GEA 1570 EUR
APOSOR32924-1G
4-Chloro-3-methanehydrazonoylquinoline, tech
4-Chloro-3-methanehydrazonoylquinoline
Formel: C₁₀H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03787776
CAS-Nummer: 866040-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR