4-Oxo-4H-benzo[h]chromene-3-carbaldehyde, tech

Lieferant: Apollo Scientific

OR32927-1G
APOSOR32927-1GEA 1570 EUR
APOSOR32927-1G
4-Oxo-4H-benzo[h]chromene-3-carbaldehyde, tech
4-Oxo-4H-benzo[h]chromene-3-carbaldehyde
Formel: C₁₄H₈O₃
Molecular Weight: 224.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787439
CAS-Nummer: 23469-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR