4-(2,3-Dihydro-1H-inden-5-yl)-4-oxobutanoic acid 95%
Lieferant: Apollo Scientific
OR32928-1G
APOSOR32928-1GEA
671
EUR
APOSOR32928-1G
4-(2,3-Dihydro-1H-inden-5-yl)-4-oxobutanoic acid 95%
4-(2,3-Dihydro-1H-inden-5-yl)-4-oxobutanoic acid
Formel:
C₁₃H₁₄O₃ Molecular Weight: 218,248 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787570 CAS-Nummer: 75382-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...