2-(3-Aminophenoxy)-1-ethanol 95%

Lieferant: Apollo Scientific

OR32930-1G
APOSOR32930-1GEA 538 EUR
APOSOR32930-1G
2-(3-Aminophenoxy)-1-ethanol 95%
2-(3-Aminophenoxy)-1-ethanol
Formel: C₈H₁₁NO₂
Molecular Weight: 153,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03004835
CAS-Nummer: 50963-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR