3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(furan-2-yl)propanoic acid, tech

Lieferant: Apollo Scientific

OR32933-1G
APOSOR32933-1GEA 1570 EUR
APOSOR32933-1G
3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(furan-2-yl)propanoic acid, tech
3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(furan-2-yl)propanoic acid
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125691
CAS-Nummer: 866040-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR