2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32935-1G
APOSOR32935-1GEA 1570 EUR
APOSOR32935-1G
2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech
2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione
Formel: C₁₆H₁₉NO₃
Molecular Weight: 273,327 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02936630
CAS-Nummer: 64869-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR