4-[4-(1H-Pyrrol-1-yl)phenyl]-1H-pyrazol-5-amine, tech

Lieferant: Apollo Scientific

OR32936-1G
APOSOR32936-1GEA 1570 EUR
APOSOR32936-1G
4-[4-(1H-Pyrrol-1-yl)phenyl]-1H-pyrazol-5-amine, tech
4-[4-(1H-Pyrrol-1-yl)phenyl]-1H-pyrazol-5-amine
Formel: C₁₃H₁₂N₄
Molecular Weight: 224,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125700
CAS-Nummer: 1024019-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR