2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid, tech

Lieferant: Apollo Scientific

OR32941-1G
APOSOR32941-1GEA 1570 EUR
APOSOR32941-1G
2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid, tech
2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid
Formel: C₁₇H₁₃NO₃
Molecular Weight: 279,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00207535
CAS-Nummer: 105576-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR