2-(1H-Pyrazol-3-yl)pyrazine 95%

Lieferant: Apollo Scientific

OR32945-250MG
APOSOR32945-250MGEA 72.5 EUR
APOSOR32945-250MG
2-(1H-Pyrazol-3-yl)pyrazine 95%
2-(1H-Pyrazol-3-yl)pyrazine
Formel: C₇H₆N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03618495
CAS-Nummer: 111781-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR