2-(5-Methyl-1H-pyrazol-3-yl)pyrazine 95%

Lieferant: Apollo Scientific

OR32946-1G
APOSOR32946-1GEA 1070 EUR
APOSOR32946-1G
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine 95%
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine
Formel: C₈H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08056640
CAS-Nummer: 192711-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR