3-[(1E)-(Hydroxyimino)methyl]-4H-chromen-4-one 95%

Lieferant: Apollo Scientific

OR32948-1G
APOSOR32948-1GEA 671 EUR
APOSOR32948-1G
3-[(1E)-(Hydroxyimino)methyl]-4H-chromen-4-one 95%
3-[(1E)-(Hydroxyimino)methyl]-4H-chromen-4-one
Formel: C₁₀H₇NO₃
Molecular Weight: 189,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00232783
CAS-Nummer: 130306-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR