2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione, tech

Lieferant: Apollo Scientific

OR32949-1G
APOSOR32949-1GEA 1570 EUR
APOSOR32949-1G
2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione, tech
2,2-Dimethyl-5-[(4-methylphenyl)methylidene]-1,3-dioxane-4,6-dione
Formel: C₁₄H₁₄O₄
Molecular Weight: 246,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128563
CAS-Nummer: 15795-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR