6-[(Pyridin-2-yl)methyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR32951-1G
APOSOR32951-1GEA 1570 EUR
APOSOR32951-1G
6-[(Pyridin-2-yl)methyl]pyridazin-3-ol, tech
6-[(Pyridin-2-yl)methyl]pyridazin-3-ol
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140707
CAS-Nummer: 338779-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR