4,5-Dichloro-6-methylpyrimidin-2-amine 95%

Lieferant: Apollo Scientific

OR32953-1G
APOSOR32953-1GEA 427 EUR
APOSOR32953-1G
4,5-Dichloro-6-methylpyrimidin-2-amine 95%
4,5-Dichloro-6-methylpyrimidin-2-amine
Formel: C₅H₅Cl₂N₃
Molecular Weight: 178,019 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244693
CAS-Nummer: 7749-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR