1-[(4-Chlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech

Lieferant: Apollo Scientific

OR32954-1G
APOSOR32954-1GEA 1570 EUR
APOSOR32954-1G
1-[(4-Chlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech
1-[(4-Chlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea
Formel: C₁₁H₁₄ClN₃O₂
Molecular Weight: 255,701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215377
CAS-Nummer: 338781-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR